Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010481.2 | NP_034611.2 | 6, 7 | 721 | ||
GenBank mRNA | AK002634.1 | - | 6, 7 | 653 | ||
AK004946.1 | - | 6, 7 | 683 | |||
AK030299.1 | - | 6, 7 | 714 | |||
AK133501.1 | - | 6, 7 | 723 | |||
AK137109.1 | - | 6, 7 | 693 | |||
AK145965.1 | - | 6, 7 | 715 | |||
AK159791.1 | - | 6, 7 | 723 | |||
AK161587.1 | - | 6, 7 | 724 | |||
AK165958.1 | - | 6, 7 | 721 | |||
AK167856.1 | - | 6, 7 | 714 | |||
AK190032.1 | - | 2, 3 | 161 | |||
AK194788.1 | - | 2, 3 | 151 | |||
AK195063.1 | - | 2, 3 | 162 | |||
BC052727.1 | - | 6, 7 | 721 | 6813991 | ||
BC057343.1 | - | 6, 7 | 721 | 6813991 | ||
D11089.2 | - | 6, 7 | 709 | |||
D17556.1 | - | 6, 7 | 693 | |||
L06896.1 | - | 6, 7 | 706 |