Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010481.2 | NP_034611.2 | 9 | 1053 | ||
GenBank mRNA | AK002634.1 | - | 9 | 985 | ||
AK004946.1 | - | 9 | 1015 | |||
AK030299.1 | - | 9 | 1046 | |||
AK133501.1 | - | 9 | 1055 | |||
AK137109.1 | - | 9 | 1025 | |||
AK145965.1 | - | 9 | 1047 | |||
AK159791.1 | - | 9 | 1055 | |||
AK161587.1 | - | 9 | 1056 | |||
AK165958.1 | - | 9 | 1053 | |||
AK167856.1 | - | 9 | 1046 | |||
BC052727.1 | - | 9 | 1053 | 6813991 | ||
BC057343.1 | - | 9 | 1053 | 6813991 | ||
D11089.2 | - | 9 | 1041 | |||
D17556.1 | - | 9 | 1025 | |||
L06896.1 | - | 9 | 1038 |