Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGB3 SIRNA Assays ›
          Gene Symbol:
          Itgb3
          Gene Name:
          integrin beta 3
          Gene Aliases:
          CD61, GP3A, INGRB3
          Chromosome Location:
          Chr.11: 104608000 - 104670490 on Build GRCm38
          UniGene ID:
          Mm.87150
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS205563
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Itgb3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016780.2 NP_058060.2 12, 13 2092
          GenBank mRNA AF026509.1 - 12, 13 2036
          AK156518.1 - 8, 9 1350
          AK157958.1 - 12, 13 2038
          BC125518.1 - 12, 13 2092 40130033
          BC125520.1 - 12, 13 2092 40130035

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          positive regulation of leukocyte migration
          protein folding
          cellular calcium ion homeostasis
          cell-substrate junction assembly
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of endothelial cell migration
          negative regulation of macrophage derived foam cell differentiation
          positive regulation of fibroblast migration
          negative regulation of lipid storage
          smooth muscle cell migration
          positive regulation of smooth muscle cell migration
          cell growth
          cell migration
          platelet activation
          regulation of cell migration
          positive regulation of cell migration
          cell-substrate adhesion
          activation of protein kinase activity
          negative regulation of lipid transport
          positive regulation of cell adhesion mediated by integrin
          positive regulation of osteoblast proliferation
          heterotypic cell-cell adhesion
          substrate adhesion-dependent cell spreading
          apolipoprotein A-I-mediated signaling pathway
          positive regulation of MAPK cascade
          positive regulation of osteoclast differentiation
          negative regulation of low-density lipoprotein particle receptor biosynthetic process
          positive regulation of angiogenesis
          positive regulation of bone resorption
          viral entry into host cell
          positive regulation of smooth muscle cell proliferation
          cell projection morphogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of lipoprotein metabolic process
          negative chemotaxis
          regulation of release of sequestered calcium ion into cytosol
          angiogenesis involved in wound healing
          negative regulation of cell death
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          positive regulation of glomerular mesangial cell proliferation
          positive regulation of adenylate cyclase-inhibiting opioid receptor signaling pathway
          fibronectin binding
          protease binding
          protein disulfide isomerase activity
          receptor activity
          integrin binding
          protein binding
          peptide binding
          identical protein binding
          vascular endothelial growth factor receptor 2 binding
          cell adhesion molecule binding
          extracellular matrix binding
          fibrinogen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0