Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145953.1 | NP_001139425.1 | 6 | 797 | ||
NM_010705.3 | NP_034835.1 | 6 | 770 | |||
GenBank mRNA | AK008593.1 | - | 5 | 439 | ||
AK089249.1 | - | 6 | 714 | |||
AK149783.1 | - | 6 | 744 | |||
AK155244.1 | - | 7 | 616 | |||
AK155484.1 | - | 6 | 712 | |||
AK159536.1 | - | 6 | 738 | |||
AK170803.1 | - | 6 | 827 | |||
BC132328.1 | - | 6 | 723 | 40130605 | ||
BC138790.1 | - | 6 | 723 | 8861812 | ||
BC145419.1 | - | 6 | 723 | 9053977 | ||
J03723.1 | - | 4 | 670 | |||
X16074.1 | - | 6 | 817 | |||
X16834.1 | - | 6 | 716 |