Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122949.2 | NP_001116421.1 | 7 | 1099 | ||
NM_001285496.1 | NP_001272425.1 | 7 | 919 | |||
NM_001285497.1 | NP_001272426.1 | 7 | 1099 | |||
NM_010823.3 | NP_034953.2 | 7 | 1075 | |||
GenBank mRNA | AF360122.1 | - | 7 | 856 | ||
AK041166.1 | - | 7 | 1047 | |||
BC101943.1 | - | 7 | 1056 | |||
BC103513.1 | - | 7 | 1032 | |||
BC103514.1 | - | 7 | 1056 | |||
BC103515.1 | - | 7 | 1056 | |||
BC146612.1 | - | 7 | 1028 | |||
BC146613.1 | - | 7 | 1033 | 40039147 | ||
X73677.1 | - | 7 | 1043 | |||
Z22649.1 | - | 7 | 1033 |