Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122949.2 | NP_001116421.1 | 3 | 343 | ||
NM_001285496.1 | NP_001272425.1 | 3 | 343 | |||
NM_001285497.1 | NP_001272426.1 | 3 | 343 | |||
NM_010823.3 | NP_034953.2 | 3 | 343 | |||
GenBank mRNA | AF360122.1 | - | 3 | 280 | ||
AK041166.1 | - | 3 | 315 | |||
BC101943.1 | - | 3 | 300 | |||
BC103513.1 | - | 3 | 300 | |||
BC103514.1 | - | 3 | 300 | |||
BC103515.1 | - | 3 | 300 | |||
BC146612.1 | - | 3 | 296 | |||
BC146613.1 | - | 3 | 301 | 40039147 | ||
Z22649.1 | - | 3 | 301 |