Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164190.1 | NP_001157662.1 | 4, 5 | 429 | ||
NM_001164191.1 | NP_001157663.1 | 4, 5 | 320 | |||
NM_001164192.1 | NP_001157664.1 | 4, 5 | 320 | |||
NM_001164193.1 | NP_001157665.1 | 4, 5 | 320 | |||
NM_019926.3 | NP_064310.2 | 4, 5 | 309 | |||
GenBank mRNA | AF073996.1 | - | 4, 5 | 310 | ||
AK016445.1 | - | 4, 5 | 430 | |||
AK031794.1 | - | 4, 5 | 291 | |||
AK032870.1 | - | 4, 5 | 273 | |||
AK149997.1 | - | 4, 5 | 322 | |||
AK166452.1 | - | 4, 5 | 288 | |||
AK171475.1 | - | 4, 5 | 298 | |||
BC051022.1 | - | 4, 5 | 278 | |||
BC066847.1 | - | 4, 5 | 267 | |||
BC090984.1 | - | 4, 5 | 287 | 30246206 |