Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285489.1 | NP_001272418.1 | 12 | 1117 | ||
NM_001285490.1 | NP_001272419.1 | 12 | 1130 | |||
NM_008672.3 | NP_032698.1 | 12 | 1117 | |||
GenBank mRNA | AJ002198.1 | - | 12 | 1087 | ||
AK090112.1 | - | 12 | 1133 | |||
AK156835.1 | - | 12 | 1090 | |||
BC089018.1 | - | 12 | 1104 | |||
BC132039.1 | - | 11 | 1041 | 40130316 | ||
BC138518.1 | - | 11 | 1041 | 8861538 | ||
BC145306.1 | - | 11 | 1041 | 9053860 | ||
BC145308.1 | - | 11 | 1041 | 9053862 |