Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164226.1 | NP_001157698.1 | 8 | 1081 | ||
NM_001164227.1 | NP_001157699.1 | 8 | 1081 | |||
NM_001164228.1 | NP_001157700.1 | 8 | 1078 | |||
NM_001164229.1 | NP_001157701.1 | 9 | 1155 | |||
NM_001164230.1 | NP_001157702.1 | 8 | 1078 | |||
NM_010938.4 | NP_035068.3 | 8 | 1078 | |||
GenBank mRNA | AF098077.1 | - | 8 | 1013 | ||
AK014494.1 | - | 8 | 1042 | |||
AK029034.1 | - | 8 | 1059 | |||
AK031103.1 | - | 3 | 282 | |||
AK037697.1 | - | 8 | 1039 | |||
AK050861.1 | - | 8 | 995 | |||
AK082580.1 | - | 8 | 1079 | |||
AK134483.1 | - | 8 | 995 | |||
AK135673.1 | - | 9 | 1158 | |||
BC005410.1 | - | 7 | 899 | 3496661 |