Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198587.3 | NP_001185516.2 | 12 | 3504 | ||
NM_172544.3 | NP_766132.2 | 11 | 2394 | |||
GenBank mRNA | AK032126.1 | - | 11 | 2386 | ||
AK129205.1 | - | 10 | 2267 | |||
BC060719.1 | - | 13 | 3079 | 6406001 |