Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009978.1 | NP_001009978.1 | 3 | 257 | ||
NM_001009979.1 | NP_001009979.1 | 3 | 257 | |||
NM_001159582.1 | NP_001153054.1 | 5 | 759 | |||
NM_016744.4 | NP_058024.2 | 4 | 554 | |||
GenBank mRNA | AF023529.1 | - | 4 | 474 | ||
AF159298.1 | - | 3 | 257 | |||
AK014810.1 | - | 3 | 256 | |||
AK043647.1 | - | 4 | 555 | |||
AK050004.1 | - | 5 | 620 | |||
AY845863.1 | - | 4 | 306 | |||
AY845864.1 | - | 3 | 257 | |||
BC090628.1 | - | 4 | 522 | 6842675 | ||
U56649.1 | - | 5 | 490 |