Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008960.2 | NP_032986.1 | 6 | 1488 | ||
GenBank mRNA | AK030750.1 | - | 2 | 251 | ||
AK076980.1 | - | 6 | 1446 | |||
AK088717.1 | - | 6 | 1467 | |||
AK148736.1 | - | 6 | 1323 | |||
BC021445.1 | - | 6 | 837 | 5038272 | ||
U92437.1 | - | 6 | 1568 |