Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138657.3 | NP_619598.1 | 8, 9 | 1710 | ||
GenBank mRNA | AF424814.1 | - | 8, 9 | 1711 | ||
AK147580.1 | - | 8, 9 | 1808 | |||
AK147650.1 | - | 8, 9 | 1808 |