Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013642.3 | NP_038670.1 | 2 | 631 | ||
GenBank mRNA | AK145903.1 | - | 2 | 636 | ||
AK149915.1 | - | 2 | 509 | |||
AK150433.1 | - | 2 | 631 | |||
AK152183.1 | - | 2 | 631 | |||
AK152324.1 | - | 2 | 634 | |||
AK152905.1 | - | 2 | 632 | |||
AK159251.1 | - | 2 | 632 | |||
AK159363.1 | - | 2 | 632 | |||
AK159567.1 | - | 2 | 632 | |||
AK159658.1 | - | 2 | 629 | |||
AK159726.1 | - | 2 | 565 | |||
AK167602.1 | - | 2 | 633 | |||
AK168627.1 | - | 2 | 632 | |||
BC006967.1 | - | 2 | 618 | 3589849 | ||
X61940.1 | - | 2 | 621 |