Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166414.1 | NP_001159886.1 | 12 | 1463 | ||
NM_011265.3 | NP_035395.2 | 11 | 1504 | |||
GenBank mRNA | AK030008.1 | - | 13 | 1536 | ||
AK041120.1 | - | 10 | 1939 | |||
AK132267.1 | - | 7 | 829 | |||
AK140928.1 | - | 10 | 1312 | |||
BC017598.1 | - | 11 | 1478 | 4483833 |