Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uimc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011307.2 | NP_035437.1 | 6 | 1080 | ||
GenBank mRNA | AK052704.1 | - | 6 | 1081 | ||
AK083342.1 | - | 6 | 1021 | |||
AK161177.1 | - | 4 | 2386 | |||
BC037092.1 | - | 6 | 1041 | |||
BC040808.1 | - | 6 | 1036 | 4038778 | ||
BC051659.1 | - | 6 | 1003 | |||
BC066220.1 | - | 6 | 1003 | |||
U22015.1 | - | 2 | 532 |