Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011403.2 | NP_035533.1 | 14 | 1995 | ||
GenBank mRNA | AK137542.1 | - | 14 | 1891 | ||
AK157181.1 | - | 14 | 1964 | |||
AK157833.1 | - | 14 | 1853 | |||
AK157861.1 | - | 14 | 1865 | |||
AK158143.1 | - | 14 | 1996 | |||
AK158184.1 | - | 14 | 1961 | |||
AK158243.1 | - | 14 | 1865 | |||
AK158453.1 | - | 14 | 1840 | |||
AK162616.1 | - | 14 | 1840 | |||
AK165195.1 | - | 14 | 1852 | |||
AK165267.1 | - | 14 | 1859 | |||
AK171662.1 | - | 1891 | ||||
AK171808.1 | - | 14 | 1829 | |||
AK171845.1 | - | 1891 | ||||
AK172103.1 | - | 14 | 1852 | |||
AK172316.1 | - | 14 | 1840 | |||
AK172395.1 | - | 14 | 1891 | |||
AK172456.1 | - | 14 | 1875 | |||
AK172569.1 | - | 14 | 1840 | |||
AY296129.1 | - | 14 | 1828 | |||
BC052419.1 | - | 14 | 1835 | 5702599 | ||
BC053429.1 | - | 14 | 1826 | 6506270 | ||
M29379.1 | - | 14 | 1822 | |||
X02677.1 | - | 14 | 1828 | |||
X03917.1 | - | 12 | 1672 |