Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205082.1 | NP_001192011.1 | 5, 6 | 932 | ||
NM_001205083.1 | NP_001192012.1 | 5, 6 | 932 | |||
NM_016741.2 | NP_058021.1 | 5, 6 | 932 | |||
GenBank mRNA | AK010622.1 | - | 5, 6 | 857 | ||
AK028191.1 | - | 5, 6 | 913 | |||
AK033114.1 | - | 5, 6 | 857 | |||
AK145588.1 | - | 5, 6 | 858 | |||
AK154933.1 | - | 5, 6 | 924 | |||
AK158122.1 | - | 5, 6 | 857 | |||
AK169906.1 | - | 5, 6 | 908 | |||
AK171602.1 | - | 5, 6 | 912 | |||
BC004656.1 | - | 5, 6 | 912 | 3498018 | ||
CT010222.1 | - | 5, 6 | 720 | |||
U37799.1 | - | 5, 6 | 770 |