Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172136.1 | NP_001165607.1 | 5, 6 | 625 | ||
NM_172456.3 | NP_766044.1 | 4, 5 | 590 | |||
GenBank mRNA | AK028887.1 | - | 4, 5 | 554 | ||
AK034654.1 | - | 5, 6 | 587 | |||
AK051300.1 | - | 5, 6 | 589 | |||
AK148426.1 | - | 4, 5 | 571 | |||
AK162566.1 | - | 4, 5 | 553 | |||
AK163555.1 | - | 4, 5 | 554 | |||
BC052171.1 | - | 3, 4 | 396 | |||
BC059920.1 | - | 5, 6 | 568 | |||
BC064730.1 | - | 5, 6 | 640 | |||
BC094589.1 | - | 2, 3 | 3076 | |||
BC119024.1 | - | 4, 5 | 554 | 8733773 | ||
BC144729.1 | - | 5, 6 | 589 |