Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113563.1 | NP_001107035.1 | 11 | 1515 | ||
NM_011489.3 | NP_035619.3 | 11 | 1847 | |||
GenBank mRNA | AK045531.1 | - | 11 | 1658 | ||
AK150098.1 | - | 11 | 1484 | |||
AK154014.1 | - | 11 | 1847 | |||
AK154664.1 | - | 11 | 1481 | |||
AY040231.1 | - | 11 | 1801 | |||
AY042906.1 | - | 11 | 1801 | |||
AY044901.1 | - | 11 | 1801 | |||
AY044902.1 | - | 11 | 1801 | |||
AY044903.1 | - | 11 | 1801 | |||
BC024319.1 | - | 11 | 1471 | 4237132 | ||
U21110.2 | - | 11 | 1801 | |||
Z48539.1 | - | 11 | 1415 |