Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem106a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144830.3 | NP_659079.1 | 9 | 901 | ||
GenBank mRNA | AK136954.1 | - | 5 | 1895 | ||
AK146019.1 | - | 9 | 902 | |||
AK149650.1 | - | 8 | 1145 | |||
AK150285.1 | - | 9 | 871 | |||
AK151842.1 | - | 9 | 877 | |||
AK152741.1 | - | 9 | 871 | |||
AK154217.1 | - | 9 | 902 | |||
AK168437.1 | - | 9 | 845 | |||
AK169081.1 | - | 9 | 844 | |||
AK170900.1 | - | 9 | 872 | |||
AK172282.1 | - | 9 | 850 | |||
BC022145.1 | - | 8 | 949 | 5101508 | ||
BC033268.1 | - | 9 | 874 | 5037750 |