Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011596.5 | NP_035726.2 | 300 | |||
GenBank mRNA | AB022323.1 | - | 266 | |||
AF218252.1 | - | 285 | ||||
AK155055.1 | - | 277 | ||||
AK170122.1 | - | 287 | ||||
BC018369.1 | - | 253 | ||||
BC108991.1 | - | 197 | 40047832 | |||
BC108992.1 | - | 197 | 40047833 | |||
BC112905.1 | - | 260 | 3670722 | |||
CJ076040.1 | - | 300 | ||||
M31226.1 | - | 196 | ||||
X55184.1 | - | 197 |