Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011650.3 | NP_035780.1 | 2 | 163 | ||
GenBank mRNA | AF234179.1 | - | 2 | 163 | ||
AK010737.1 | - | 2 | 161 | |||
AK033003.1 | - | 1 | 403 | |||
AK150529.1 | - | 2 | 212 | |||
AK151448.1 | - | 2 | 214 | |||
AK165598.1 | - | 2 | 203 | |||
AK181409.1 | - | 2 | 129 | |||
AK183078.1 | - | 2 | 129 | |||
AK184665.1 | - | 2 | 133 | |||
AK198107.1 | - | 2 | 129 | |||
AK199692.1 | - | 2 | 129 | |||
AK202525.1 | - | 2 | 129 | |||
BC004615.1 | - | 2 | 209 | 3585366 | ||
X81464.1 | - | 2 | 163 |