Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009441.2 | NP_033467.2 | 23 | 2183 | ||
GenBank mRNA | AB008516.1 | - | 22 | 2060 | ||
AK053765.1 | - | 23 | 2287 | |||
AK084201.1 | - | 13 | 1218 | |||
AK088273.1 | - | 22 | 2130 | |||
AK165850.1 | - | 12 | 1091 | |||
AK166120.1 | - | 13 | 1123 | |||
AK166164.1 | - | 14 | 1272 | |||
AK220495.1 | - | 23 | 2225 | |||
BC019173.1 | - | 23 | 2152 | |||
BC037654.1 | - | 22 | 2083 | |||
BC057207.1 | - | 22 | 2087 | |||
BC145489.1 | - | 18 | 1703 |