Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009466.2 | NP_033492.1 | 10, 11 | 1486 | ||
GenBank mRNA | AF061017.1 | - | 10, 11 | 1370 | ||
AK144673.1 | - | 10, 11 | 1522 | |||
AK146693.1 | - | 10, 11 | 1370 | |||
AK149590.1 | - | 4, 5 | 2766 | |||
AK162298.1 | - | 10, 11 | 1374 | |||
AK167462.1 | - | 10, 11 | 1366 | |||
AK167560.1 | - | 10, 11 | 1466 | |||
AK168064.1 | - | 8, 9 | 1083 | |||
AK168508.1 | - | 10, 11 | 1373 | |||
AK168563.1 | - | 10, 11 | 1370 | |||
AK169208.1 | - | 10, 11 | 1501 | |||
AK190373.1 | - | 1, 2 | 101 | |||
BC006749.1 | - | 10, 11 | 1486 | 3708709 |