Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009511.2 | NP_033537.1 | 13 | 1352 | ||
GenBank mRNA | BC138572.1 | - | 13 | 1250 | 8861592 | |
D28132.1 | - | 13 | 1284 |
Set Membership: |
![]() |