Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb561a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282064.1 | NP_001268993.1 | 4 | 1065 | ||
NM_001282065.1 | NP_001268994.1 | 3 | 1162 | |||
NM_001282067.1 | NP_001268996.1 | 5 | 1085 | |||
NM_201351.2 | NP_958739.1 | 4 | 1255 | |||
GenBank mRNA | AK040692.1 | - | 959 | |||
AK139533.1 | - | 4 | 1243 | |||
AK160179.1 | - | 3 | 866 | |||
AK168841.1 | - | 4 | 959 | |||
AK169729.1 | - | 4 | 961 | |||
AK170911.1 | - | 5 | 753 | |||
AK172638.1 | - | 4 | 772 | |||
BC065078.1 | - | 4 | 689 | 6817093 |