Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113416.1 | NP_001106887.1 | 5, 6 | 633 | ||
NM_145506.4 | NP_663481.2 | 5, 6 | 633 | |||
GenBank mRNA | AK034676.1 | - | 5, 6 | 555 | ||
AK037024.1 | - | 5, 6 | 589 | |||
AK040354.1 | - | 5, 6 | 601 | |||
AK044959.1 | - | 5, 6 | 571 | |||
AK044996.1 | - | 5, 6 | 579 | |||
AK047523.1 | - | 5, 6 | 560 | |||
AK079301.1 | - | 5, 6 | 584 | |||
AK136125.1 | - | 5, 6 | 588 | |||
AK161141.1 | - | 5, 6 | 584 | |||
AK167449.1 | - | 5, 6 | 633 | |||
AK173198.1 | - | 5, 6 | 567 | |||
BC003937.1 | - | 5, 6 | 618 | 3486543 | ||
BC011476.1 | - | 5, 6 | 567 | 4014009 |