Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178690.4 | NP_848805.2 | 21 | 2487 | ||
GenBank mRNA | AK076244.1 | - | 21 | 2451 | ||
AK076399.1 | - | 21 | 2470 | |||
AK080432.1 | - | 21 | 2469 | |||
AK134604.1 | - | 21 | 2469 | |||
AK164523.1 | - | 21 | 2468 | |||
AK170768.1 | - | 21 | 2460 | |||
AK172881.1 | - | 21 | 2474 | |||
AK194063.1 | - | 2 | 126 | |||
BC028996.1 | - | 5 | 895 | |||
BC038485.1 | - | 5 | 801 | |||
BC046297.1 | - | 21 | 2453 | 6486170 |