Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310035C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029349.1 | NP_083625.1 | 9, 10 | 1801 | ||
NM_173187.3 | NP_775279.2 | 10, 11 | 1873 | |||
GenBank mRNA | AK035724.1 | - | 9, 10 | 1783 | ||
AK040168.1 | - | 11, 12 | 1509 | |||
AK050268.1 | - | 3, 4 | 362 | |||
AK129368.1 | - | 9, 10 | 1774 | |||
AK148005.1 | - | 10, 11 | 1853 | |||
BC060194.1 | - | 9, 10 | 1046 | |||
BC117949.1 | - | 10, 11 | 1807 | 40098078 | ||
BC117950.1 | - | 9, 10 | 1739 | 40098082 |