Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310035C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029349.1 | NP_083625.1 | 15 | 2507 | ||
NM_173187.3 | NP_775279.2 | 16 | 2579 | |||
GenBank mRNA | AK035724.1 | - | 15 | 2489 | ||
AK040168.1 | - | 17 | 2215 | |||
AK050268.1 | - | 9 | 1069 | |||
AK129368.1 | - | 15 | 2480 | |||
AK148005.1 | - | 16 | 2559 | |||
BC060194.1 | - | 15 | 1752 | |||
BC117949.1 | - | 16 | 2513 | 40098078 | ||
BC117950.1 | - | 15 | 2445 | 40098082 |