Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178693.4 | NP_848808.1 | 5 | 449 | ||
GenBank mRNA | AK039422.1 | - | 5 | 450 | ||
AK039676.1 | - | 5 | 450 | |||
AK043574.1 | - | 5 | 425 | |||
AK050459.1 | - | 5 | 459 | |||
AK163270.1 | - | 1189 | ||||
BC139373.1 | - | 5 | 492 | 8862395 | ||
BC139376.1 | - | 5 | 489 | 8862398 | ||
BC145615.1 | - | 5 | 463 |