Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252584.2 | NP_001239513.1 | 11 | 1599 | ||
NM_001252585.1 | NP_001239514.1 | 10 | 1491 | |||
NM_001252587.2 | NP_001239516.1 | 12 | 1611 | |||
NM_001281926.1 | NP_001268855.1 | 11 | 1498 | |||
NM_001291158.1 | NP_001278087.1 | 12 | 3608 | |||
NM_027230.5 | NP_081506.3 | 12 | 1686 | |||
GenBank mRNA | AB093284.2 | - | 12 | 3600 | ||
AK132427.1 | - | 10 | 1493 | |||
AK147620.1 | - | 11 | 1499 | |||
AK155864.1 | - | 11 | 1404 | |||
BC020059.1 | - | 1 | 411 | |||
BC023300.1 | - | 11 | 1466 | |||
BC041797.1 | - | 1 | 414 | |||
BC048186.1 | - | 12 | 1493 | 6827541 | ||
BC063267.1 | - | 11 | 1392 |