Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252584.2 | NP_001239513.1 | 11 | 1386 | ||
NM_001252585.1 | NP_001239514.1 | 10 | 1278 | |||
NM_001252587.2 | NP_001239516.1 | 12 | 1398 | |||
NM_001281926.1 | NP_001268855.1 | 11 | 1285 | |||
NM_001291158.1 | NP_001278087.1 | 12 | 3395 | |||
NM_027230.5 | NP_081506.3 | 12 | 1473 | |||
GenBank mRNA | AB093284.2 | - | 12 | 3387 | ||
AK132427.1 | - | 10 | 1280 | |||
AK147620.1 | - | 11 | 1286 | |||
AK155864.1 | - | 11 | 1191 | |||
BC020059.1 | - | 1 | 198 | |||
BC023300.1 | - | 11 | 1253 | |||
BC041797.1 | - | 1 | 201 | |||
BC048186.1 | - | 12 | 1280 | 6827541 | ||
BC063267.1 | - | 11 | 1179 |