Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194344.2 | NP_919325.2 | 12 | 2142 | ||
GenBank mRNA | AK028482.1 | - | 8 | 2238 | ||
AK131143.1 | - | 6 | 1007 | |||
AK150064.1 | - | 12 | 1920 | |||
AK155330.1 | - | 12 | 1907 | |||
AK159089.1 | - | 12 | 1921 | |||
AK167283.1 | - | 12 | 1843 | |||
AK170383.1 | - | 12 | 1975 | |||
AK170396.1 | - | 12 | 1922 | |||
AK171209.1 | - | 12 | 1899 | |||
AK171242.1 | - | 12 | 1899 | |||
BC024909.1 | - | 10 | 1592 | |||
BC056461.1 | - | 10 | 1486 |