Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172733.1 | NP_766321.1 | 6 | 551 | ||
GenBank mRNA | AK012558.1 | - | 6 | 587 | ||
AK034347.1 | - | 553 | ||||
AK075963.1 | - | 6 | 576 | |||
AK083298.1 | - | 6 | 551 | |||
AK145820.1 | - | 6 | 587 | |||
AK150273.1 | - | 6 | 577 | |||
AK150712.1 | - | 6 | 547 | |||
AK152462.1 | - | 6 | 555 | |||
AK168204.1 | - | 6 | 587 | |||
AK169418.1 | - | 6 | 571 | |||
AK179075.1 | - | 2 | 56 | |||
BC016218.1 | - | 6 | 557 | 4484163 |