Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172733.1 | NP_766321.1 | 2 | 145 | ||
GenBank mRNA | AK012558.1 | - | 2 | 181 | ||
AK030961.1 | - | 2 | 177 | |||
AK034347.1 | - | 147 | ||||
AK075963.1 | - | 2 | 170 | |||
AK083298.1 | - | 2 | 145 | |||
AK145820.1 | - | 2 | 181 | |||
AK150273.1 | - | 2 | 171 | |||
AK150712.1 | - | 2 | 141 | |||
AK152462.1 | - | 2 | 149 | |||
AK168204.1 | - | 2 | 181 | |||
AK169418.1 | - | 2 | 165 | |||
BC016218.1 | - | 2 | 151 | 4484163 |