Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172772.2 | NP_766360.2 | 1 | 786 | ||
GenBank mRNA | AK035930.1 | - | 1 | 787 | ||
AK046403.1 | - | 1 | 773 | |||
AK046623.1 | - | 1 | 760 | |||
AK220158.1 | - | 1 | 815 | |||
BC049357.1 | - | 1 | 763 | |||
BC055816.1 | - | 1 | 514 | |||
BC058683.1 | - | 1 | 744 | 6834871 | ||
BC069845.1 | - | 1 | 499 |
Gene Symbol: Gm10642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK159631.1 | - | 1 | 136 |