Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172772.2 | NP_766360.2 | 8 | 1655 | ||
GenBank mRNA | AK035930.1 | - | 8 | 1656 | ||
AK046403.1 | - | 8 | 1642 | |||
AK077470.1 | - | 5 | 587 | |||
AK186355.1 | - | 2 | 116 | |||
AK214468.1 | - | 2 | 106 | |||
AK214619.1 | - | 3 | 323 | |||
AK216605.1 | - | 3 | 323 | |||
AK220158.1 | - | 8 | 1684 | |||
BC049357.1 | - | 8 | 1632 | |||
BC055816.1 | - | 8 | 1383 | |||
BC058683.1 | - | 8 | 1613 | 6834871 | ||
BC069845.1 | - | 8 | 1368 |