Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172772.2 | NP_766360.2 | 7, 8 | 1600 | ||
GenBank mRNA | AK035930.1 | - | 7, 8 | 1601 | ||
AK046403.1 | - | 7, 8 | 1587 | |||
AK077470.1 | - | 4, 5 | 532 | |||
AK212033.1 | - | 3, 4 | 382 | |||
AK214468.1 | - | 1, 2 | 51 | |||
AK216605.1 | - | 2, 3 | 268 | |||
AK220158.1 | - | 7, 8 | 1629 | |||
BC049357.1 | - | 7, 8 | 1577 | |||
BC055816.1 | - | 7, 8 | 1328 | |||
BC058683.1 | - | 7, 8 | 1558 | 6834871 | ||
BC069845.1 | - | 7, 8 | 1313 |