Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011875.4 | NP_036005.1 | 3, 4 | 279 | ||
GenBank mRNA | AF107838.1 | - | 3, 4 | 240 | ||
AK051078.1 | - | 3, 4 | 270 | |||
AK075936.1 | - | 3, 4 | 280 | |||
AK081160.1 | - | 3, 4 | 277 | |||
AK145654.1 | - | 3, 4 | 242 | |||
AK163017.1 | - | 3, 4 | 274 | |||
AK167169.1 | - | 3, 4 | 264 | |||
AK171504.1 | - | 3, 4 | 260 | |||
AK206994.1 | - | 3, 4 | 155 | |||
BC029989.1 | - | 3, 4 | 219 | |||
BC139184.1 | - | 3, 4 | 257 | 8862206 | ||
BC139187.1 | - | 3, 4 | 257 | 8862209 |