Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026496.4 | NP_080772.2 | 12, 13 | 1872 | ||
GenBank mRNA | AF411213.1 | - | 12, 13 | 1605 | ||
BC004783.1 | - | 3, 4 | 278 | |||
BC042575.1 | - | 7, 8 | 705 | |||
BC055035.1 | - | 12, 13 | 1661 | 6306106 |