Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146937.1 | NP_667148.1 | 1 | 633 | ||
GenBank mRNA | BC127997.1 | - | 1 | 637 | 40135782 |
Set Membership: |
![]() |