Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291787.1 | NP_001278716.1 | 22 | 1603 | ||
NM_009006.3 | NP_033032.1 | 22 | 1629 | |||
GenBank mRNA | AK011708.1 | - | 20 | 1486 | ||
AK042262.1 | - | 20 | 1351 | |||
AK051036.1 | - | 21 | 1418 | |||
AK078526.1 | - | 13 | 857 | |||
AK154164.1 | - | 19 | 2074 | |||
AK154323.1 | - | 22 | 1534 | |||
AK216809.1 | - | 4 | 223 | |||
BC116286.1 | - | 21 | 1607 | 40094503 | ||
BC116287.1 | - | 22 | 1486 | 40094508 | ||
U50595.1 | - | 22 | 1539 |