Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290654.1 | NP_001277583.1 | 2 | 435 | ||
NM_001290655.1 | NP_001277584.1 | 2 | 440 | |||
NM_172391.4 | NP_765979.3 | 2 | 440 | |||
GenBank mRNA | AK033696.1 | - | 2 | 348 | ||
AK035941.1 | - | 2 | 213 | |||
AK093945.1 | - | 2 | 347 | |||
AK137116.1 | - | 2 | 351 | |||
AK154196.1 | - | 2 | 351 | |||
AK155265.1 | - | 2 | 351 | |||
BC038397.1 | - | 2 | 326 | 3676293 | ||
BC052829.1 | - | 2 | 358 | 30040034 | ||
BC055873.1 | - | 2 | 302 | |||
BC064012.1 | - | 2 | 260 | 30135289 | ||
CJ147821.1 | - | 2 | 441 |