Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178746.4 | NP_848861.1 | 13 | 1550 | ||
GenBank mRNA | AK043402.1 | - | 13 | 1538 | ||
AK044735.1 | - | 13 | 1540 | |||
AK078926.1 | - | 7 | 790 |