Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177806.1 | NP_001171277.1 | 37 | 4570 | ||
NM_013719.3 | NP_038747.2 | 38 | 4874 | |||
GenBank mRNA | AJ243533.1 | - | 38 | 4874 | ||
AK011380.1 | - | 11 | 960 | |||
AK129334.1 | - | 33 | 4224 | |||
BC072637.1 | - | 37 | 4570 | 30354433 | ||
BM250244.2 | - | 2 | 335 |
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156845.1 | - | 2525 |