Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177806.1 | NP_001171277.1 | 38 | 4717 | ||
NM_013719.3 | NP_038747.2 | 39 | 5021 | |||
GenBank mRNA | AF193342.1 | - | 34 | 4924 | ||
AF193343.1 | - | 39 | 5013 | |||
AF193344.1 | - | 39 | 5105 | |||
AI324072.1 | - | 1 | 196 | |||
AJ243533.1 | - | 39 | 5021 | |||
AK011380.1 | - | 12 | 1107 | |||
AK129334.1 | - | 34 | 4371 | |||
BC072637.1 | - | 38 | 4717 | 30354433 | ||
BM250244.2 | - | 1 | 188 |
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156845.1 | - | 1531 |