Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013876.3 | NP_038904.1 | 2 | 539 | ||
GenBank mRNA | AB024427.1 | - | 3 | 404 | ||
AF220206.1 | - | 2 | 293 | |||
AK014254.1 | - | 2 | 693 | |||
AK183757.1 | - | 1 | 154 | |||
AK210997.1 | - | 1 | 154 | |||
BC010299.1 | - | 2 | 539 | 3155804 | ||
BC028255.1 | - | 2 | 720 | 4972336 |