Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177185.4 | NP_796159.3 | 15 | 3882 | ||
GenBank mRNA | AK051609.1 | - | 15 | 3255 | ||
AK173339.1 | - | 3 | 4918 | |||
BC051458.1 | - | 15 | 3854 | 6468017 |